Semi-Micro Cuvettes, Fireflysci
Marketsource
Standard Semi-micro spectrophotometer cuvette with PTFE Cover.
- Gives highly accurate results, only need to test sample once
- Comes in several different materials: Optical Glass, UV Quartz, IR Quartz
- Excellent transmission values
- Fire-fused to ensure that cuvettes are robust and durable
Delivery information: Lightpaths from 5 – 100 mm are available.
Standard Semi-micro spectrophotometer cuvette with PTFE Cover.
- Gives highly accurate results, only need to test sample once
- Comes in several different materials: Optical Glass, UV Quartz, IR Quartz
- Excellent transmission values
- Fire-fused to ensure that cuvettes are robust and durable
Delivery information: Lightpaths from 5 – 100 mm are available.
Product Family Options
- DescriptionCapacityOuter heightOuter widthLightpathCover type
- : 9UV5
- : 9UV10
- : 9UV20MarketsourceMarketsource
- : 9UV30MarketsourceMarketsource
- : 9UV40MarketsourceMarketsource
- : 9UV50MarketsourceMarketsource
- Cuvette, Outer Length: 102.5 mm14.000 ml45 mm12.5 mm100 mmPTFEEach$1,195.33: 9UV100MarketsourceMarketsource