DKK-1 ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-386
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Primary antibody reactivity:Human
- Target protein:Dkk-1
- Description:DKK-1 ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-386
Specifications
About this item
Human DKK-1 ELISA is to detect Human DKK-1 in serum.
- Dynamic Range: 1.56-50 ng/ml
- Sensitivity: 0.6 ng/ml
- Incubation: 1h40
Dickkopf-1(DKK-1) is a 28.67 kDa secreted protein that acts as soluble inhibitor of the WNT signalling pathway. This pathway contains lipid-modified glycoproteins that activate cell surface receptor-mediated signal transduction to regulate cell activities like: cell fate, proliferation, migration, polarity and gene expression. DKK-1 regulates developmental processes of all kinds. Thus, DKK-1 is also involved in the regulation of bone metabolism as it effects osteoblast differentiation and in regulation of tumourgenetic activity.