Etanercept ELISA (mAb-based) ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-222
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Description:Etanercept ELISA (mAb-based) ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-222
Specifications
About this item
Etanercept ELISA is to measure of Etanercept in serum and plasma samples.
- Dynamic Range: 0.025 - 10 ng/ml
- Sensitivity: 0.025 ng/ml
- Incubation: 1.5 hours
The Eagle Biosciences Etanercept is an enzyme immunoassay for the specific and precise measurement of free Etanercept in serum and plasma samples. The Etanercept ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.