Order Entry
Canada
ContactUsLinkComponent
CoproELISA H. pylori, Eagle Biosciences
CoproELISA H. pylori, Eagle Biosciences
Catalog # 103389-352
Supplier:  Eagle Biosciences
CAS Number:  
CoproELISA H. pylori, Eagle Biosciences
Catalog # 103389-352
Supplier:  Eagle Biosciences
Supplier Number:  774-01
CAS Number:  

Specifications

  • Description:
    CoproELISA H. pylori
  • Size:
    1 kit
  • Cat. no.:
    103389-352

Specifications

About this item

CoproELISA Entamoeba detects Entamoeba antigens in unpreserved human fecal specimens.

  • Dynamic Range: Cut-Off
  • Sensitivity: Cut-Off
  • Incubation: 2 hours

Helicobacter pylori (H. pylori) is a Gram-negative, micro-aerophilic  bacterium restricted to the stomach and the duodenum. It was identified in 1982 by Australian scientists Barry Marshall and Robin Warren, who found that it was present in patients with chronic gastritis and gastric ulcers, conditions that were not previously believed to have a microbial cause. H. pylori has also been associated as an etiologic agent in the development of duodenal ulcers and stomach cancer. More than fifty percent of the world's population harbor H. pylori in their upper gastrointestinal tract. However, over 80 percent of individuals infected with this bacterium are asymptomatic and it has been postulated that it may play an important role in the natural stomach ecology.  H. pylori is a contagious bacterium, although the exact route of transmission is not known. Person-to-person transmission by either the oral-oral or fecal-oral route is most frequent. Consistent with these transmission routes, the bacteria have been isolated from feces, saliva and dental plaques of some infected patients. Transmission occurs mainly within families in developed nations, yet can also be acquired from the community in developing countries.