Order Entry
Canada
ContactUsLinkComponent
Aflibercept (Eyelea®, Zaltrap®) ELISA Assay Kit, Eagle Biosciences, Inc.
Aflibercept (Eyelea®, Zaltrap®) ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 103389-218
Supplier:  Eagle Biosciences
CAS Number:  
Aflibercept (Eyelea®, Zaltrap®) ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 103389-218
Supplier:  Eagle Biosciences
Supplier Number:  IG-AA115
CAS Number:  

Specifications

  • Target protein:
    Aflibercept (Eyelea®, Zaltrap®)
  • Description:
    Aflibercept (Eyelea®, Zaltrap®) ELISA Assay Kit
  • Size:
    1 kit
  • Cat. no.:
    103389-218

Specifications

About this item

Aflibercept ELISA is intended to measure free Aflibercept in serum and plasma samples.

  • Dynamic Range: 6 - 200 ng/mL
  • Sensitivity: 5 ng/mL
  • Incubation: 2 hours

The drug Aflibercept (trade names Eylea®, Zaltrap®) is a recombinant fusion protein consisting of VEGF-binding portions from the extracellular domains of human VEGF receptors 1 and 2 fused to the Fc portion of human IgG1. Aflibercept has broad affinity for all ligands that bind to these receptors, including isoforms of VEGF-A, VEGF-B, and placental growth factors. Aflibercept demonstrated antitumour effects and antiangiogenic activity as a single agent and enhanced activity in combination with chemotherapy. Identification of biomarkers for (non-)response and risk factors for adverse drug reactions that might be related to serum concentrations and maintaining the effective concentration of Aflibercept in order to potentially avoid some side effects with a reliable method might be beneficial.