Mouse / Rat Dopamine ELISA, Eagle Biosciences
Catalog # 103389-154
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Primary antibody reactivity:Mouse
- Description:Mouse / Rat Dopamine ELISA
- Size:1 kit
- Cat. no.:103389-154
Specifications
About this item
Mouse/Rat Dopamine ELISA detects dopamine in serum, plasma, tissue, and cell culture samples.
- Dynamic Range: 0.3 - 100 ng/mL
- Sensitivity: 5.9 pg/mL
- Incubation: overnight
The Mouse/Rat Dopamine ELISA Assay kit is an Enzyme-linked immunosorbent assay used for the quantitative and very sensitive determination of noradrenaline in biological samples including serum, plasma, tissue, and cell culture samples. The Mouse/Rat Dopamine ELISA Assay kit is for research use only and should not be used in diagnostic procedures.