Order Entry
Canada
ContactUsLinkComponent
MMP-13 ELISA Assay Kit, Eagle Biosciences
MMP-13 ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-410
Supplier:  Eagle Biosciences
CAS Number:  
MMP-13 ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-410
Supplier:  Eagle Biosciences
Supplier Number:  30518101
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Conjugate ELISA:
    HRP-Streptavidin
  • Target protein:
    MMP-13
  • Description:
    MMP-13 ELISA Assay Kit
  • Size:
    1 kit
  • Detection range:
    32 - 2000 pg/ml
  • Sample volume:
    10 µl
  • Sensitivity:
    7 pg/ml
  • Regulatory status:
    RUO
  • Cat. no.:
    103389-410

Specifications

About this item

MMP-13 ELISA is sensitive and specific to determine activated human MMP-13 in biological samples.

  • Dynamic Range: 32 -2000 pg/ml
  • Sensitivity: 7 pg/ml
  • Incubation: 4.25 hours

Matrix Metalloproteinases (MMPs) comprise a family of secreted and membrane-bound endopeptidases which hydrolyse extracellular matrix proteins (1). Based on their preferred substrates and on structural features MMPs can be divided into collagenases, gelatinases, stromelysins and membrane-type matrix metalloproteinases. Their activity is tightly regulated by inhibitors (2). Collagenase 3 (MMP-13) is a secreted 452 residue protein which is released from cells as an inactive zymogen and activated extracellularly by removal of its propeptide. Cleavage of the 84 residue propeptide can be catalysed by other MMPs such as MMP-2 and MMP-14, or by other proteases like plasmin (3). Under normal conditions, MMP-13 is expressed during embryogenesis (fetal bone development) and is present only at very low levels in adult tissue. However, this enzyme is reported to be involved in the development and metastasis of breast and lung carcinomas, chondrosarcomas and osteosarcomas, head and neck carcinomas and some forms of skin cancer (4). Additionally, this enzyme plays an important role in degenerative bone diseases including osteoarthritis (5) and rheumatoid arthritis (6). Our MMP-13 test kit enables the user to measure the concentration of this putative marker enzyme in body fluids such as serum and synovial fluid with high sensitivity. The ELISA MMP-13 is specific for the activated enzyme.