Mouse / Rat Adrenaline (Epinephrine) High Sensitive ELISA, Eagle Biosciences
Catalog # 103389-146
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Primary antibody reactivity:Mouse
- Description:Mouse / Rat Adrenaline (Epinephrine) High Sensitive ELISA
- Size:1 kit
- Cat. no.:103389-146
Specifications
About this item
Adrenaline Sensitive ELISA is intended to detect Adrenaline in biological fluids by a microplate .
- Dynamic Range: 0.15 - 25 ng/ml
- Sensitivity: 1.6 pg/mL
- Incubation: overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.