Specifications
- Description:SeroCT RT C. trachomatis IgG ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-318
Specifications
About this item
SeroCT RT IgG ELISA is intended to detect IgG antibodies specific to C.trachomatis in human serum.
- Dynamic Range: Cut-off
- Sensitivity: Cut-off
- Incubation: 1.5 hours
Chlamydia is a gram-negative obligate intracellular bacteria that causes acute and chronic diseases in mammalian and avian species. The genus Chlamydia is comprised of four species: C.trachomatis, C.pneumoniae, C.psittaci and C. pecorum. C.trachomatis is divided into 15 serovars (5-8). Serovars A, B, Ba and C are agents of trachoma, the leading cause of preventable blindness endemic in third world countries. Serovars L1-L3 are the agents of lymphogranuloma venereum. Serovars D-K are the common cause of sexually transmitted genital infection worldwide: cervicitis, endometritis/ salpingitis (10) in females and urethritis (11) in both males and females. Endometritis/salpingitis can lead to tubal occlusion with a higher risk of extrauterine pregnancy and infertility. Genital infection may cause an acute and persistent infection occasionally without any clinical symptoms. Generally, these infections are treatable once they are diagnosed. However without any treatment the infection may progress to a severe chronic inflammation leading to infertility, ectopic pregnancy, induced abortion or child delivery. Moreover, infants to infected mothers may be infected during birth, leading to conjunctivitis or pneumonia. The serology of C. trachomatis is more interesting in cases of chronic infections than in acute infections. C.pneumoniae is an important respiratory pathogen in humans and causes up to 10% of community-acquired pneumonia cases. It has been associated with acute respiratory diseases, pneumonia, asthma, bronchitis, pharyngitis, acute chest syndrome of sickle cell disease, coronary heart disease, and Guillain-Barre syndrome. C.psittaci infects a diverse range of host species from molluscs to birds to mammals and also causes severe pneumonia. In animals, C.psittaci and C.pecorum are capable of inducing diverse disease syndromes like pneumonia, enteritis, polyserositis, encephalitis and conjunctivitis. Serological testing, now an established approach in many countries, has been shown to provide a comprehensive answer to detect C. trachomatis infection. In suspected deep-seated infection, serum sampling reduces the necessity for invasive procedures, which are required for direct antigen detection. In cases of lower urogenital infections, collection limitations such as effectiveness of scrape sampling procedure, specimen handling and transportation difficulties have to be weighed. Above all, the issue remains that most Chlamydia infections are asymptomatic. Therefore an infection may persist for a long time, ascend the upper genital tract, causing deep and chronic infections, and increase the probability of false negative results in direct antigen detection. Serological testing for Chlamydia trachomatis, through the detection of various specific antibodies, is today an effective and highly accepted option. New and accurate technologies apply the immuno markers IgM, IgA and IgG to characterize the presence and stage of infection. Specific IgM is indicative of acute Chlamydia infections. Absence does not; however preclude the presence of on-going infection, especially in recurrent and chronic cases. The use of specific IgA as a marker for active Chlamydia infection has been shown to have an important role because of its short half-life time, while persisting as long as antigenic stimulation exists. IgA, however, is more suitable for post therapy follow-up. IgG is a marker for Chlamydia positive immune response in either current, chronic or past infections. Serological cross-reactions occur between the three different species of Chlamydia. Most of the serological diagnostic assays for Chlamydia use either purified elementary bodies microimmunofluorescence (MIF) and ELISA tests, lipopolysaccharides (LPS) or purified major outer membrane protein (MOMP), as antigens. Genus specific epitopes are present in all the above antigens, therefore low species specificity is observed. Moreover, a large proportion of the population has been exposed to C.pneumoniae (with no clinical signs), the prevalence of anti- Chlamydia antibodies is very high. Therefore, the differentiation between C.pneumoniae and C. trachomatis specific antibodies using conventional serological screening tests (MIF, ELISA, EIA etc.) is insufficient. Savyon® Diagnostics Ltd has developed an assay in which C.trachomatis species specific epitopes, derived from different serotypes, are used in an Enzyme-Linked Immunosorbent Assay (ELISA). The test excludes cross-species reactive epitopes and enables more accurate and more specific determination of C.trachomatis IgG and IgA antibodies.