Rituximab ELISA (mAb-based) ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 103389-228
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Description:Rituximab ELISA (mAb-based) ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-228
Specifications
About this item
Rituximab ELISA is a mAb-based that detects the concentration of Rituximab in serum and plasma.
- Dynamic Range: 6 - 200 ng/mL
- Sensitivity: 2 ng/mL
- Incubation: 2 hours
The Eagle Biosciences Rituximab (Rituxan®, Mabthera®) ELISA Assay Kit is developed for specific measurement of Rituximab in serum and plasma. The kit is a solid phase enzyme-linked immunosorbent assay (ELISA) is based on rituximab-specific 9F9 monoclonal antibody (mAb). The Eagle Biosciences Rituximab (Rituxan®, Mabthera®) ELISA Assay Kit is for research use only and is not intended for diagnostic or therapeutic procedures.