Fecal C. Difficile Toxin B Qualitative ELISA Assay Kit, Eagle Biosciences, Inc.
Catalog # 103389-164
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Description:Fecal C. Difficile Toxin B Qualitative ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-164
Specifications
About this item
Clostridium Difficile Toxin B ELISA is intended for qualitative detection of C
- Dynamic Range: Cut-off
- Sensitivity: Cut-off
- Incubation: 2 hours
Clostridium difficile is a common pathogen and a major cause of infectious diarrhea in hospitalized patients. While most strains produce both Toxin A and Toxin B, some only produce one or the other. These enterotoxins, which are both proinflamatory and cytotoxic, attack the mucosal lining of the intestines. If not identified and treated in a timely fashion, can result in permanent damage to the colon or colitis. This assay employs an antibody specific to Toxin B without cross-reactivity to toxin A.
Difficile Toxin B.