Antibody to Golimumab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-252
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Description:Antibody to Golimumab ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-252
Specifications
About this item
Antibody to Golimumab (Simponi) ELISA determines free antibodies to Golimumab in serum and plasma.
- Dynamic Range: Cut-off
- Sensitivity: Cut-off
- Incubation: 2.5 hours
The Eagle Biosciences Antibody to Golimumab ELISA Assay Kit is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Golimumab in serum and plasma samples. The Antibody to Golimumab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.