Order Entry
Canada
ContactUsLinkComponent
Functional Leptin ELISA, Eagle Biosciences, Inc.
Functional Leptin ELISA, Eagle Biosciences, Inc.
Catalog # 103389-282
Supplier:  Eagle Biosciences
CAS Number:  
Functional Leptin ELISA, Eagle Biosciences, Inc.
Catalog # 103389-282
Supplier:  Eagle Biosciences
Supplier Number:  L07
CAS Number:  

Specifications

  • Target protein:
    Leptin
  • Description:
    Functional Leptin ELISA
  • Size:
    1 kit
  • Cat. no.:
    103389-282

Specifications

About this item

Functional Leptin ELISA measures the functional, receptor binding Leptin levels in serum or plasma.

  • Dynamic Range: 0.05 - 6 ng/mL
  • Sensitivity: 0.01 ng/ml
  • Incubation: 4 hours

Leptin a hormone of 146 amino acids shows a molecular weight of 16 kDa (P41159). It consists of four anti-parallel a-helices. Leptin is primarily produced by adipocytes and thus provides a signal to the energy state of the organism. By binding to the leptin receptor (P48357) it influences the activity of the JAK -STAT pathway and thereby regulates energy metabolism, especially the food intake. Leptin levels show a circadian variation (+/- 30%) and are dependent on BMI, pubertal status, and gender. In addition to the expression itself, the bioavailability of leptin, is regulated via a binding protein, as with many hormones with type I cytokine receptors. The binding protein is the extracellular domain of the Leptin receptor formed by proteolytic cleavage of the receptor be metalloproteases (ADAM 10/17).