Order Entry
Canada
ContactUsLinkComponent
Human VDBP (GC Globulin) ELISA Assay Kit, Eagle Biosciences
Human VDBP (GC Globulin) ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-182
Supplier:  Eagle Biosciences
CAS Number:  
Human VDBP (GC Globulin) ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-182
Supplier:  Eagle Biosciences
Supplier Number:  DBP31-K01
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human VDBP (GC Globulin) ELISA Assay Kit
  • Size:
    1 kit
  • Cat. no.:
    103389-182

Specifications

About this item

Human VDBP (GC Globulin) ELISA determines free Vitamin D Binding Protein in serum and plasma.

  • Dynamic Range: 0 - 20.1 µg/mL
  • Sensitivity: 0.0637 µg/mL
  • Incubation: 1.5 hours

Vitamin D-binding protein (DBP) is a 58 kDa circulating glycoprotein secreted by the liver. It binds about 85% to 90% of circulating 25-OH-D2/3 and 1,25-(OH)2-D3, regulates the bioavailability of active vitamin D, and transports them to target organs. There is only less than one percent 25-OH-D2/3 is in the free form in the circulation. The full length DBP contains 476-amino acids, including a 16-amino acid signal sequence. Biologically, DBP may involve directly and indirectly in regulating bone metabolism. Circulating DBP also binds to actin at 1:1 molecule ratio, while this protein complex is removed by kidney. In patient with trauma, sepsis or multiple organ failure, DBP concentration decreases. Urine DBP is reported to be a biomarker of major renal event in patient undergoing coronary angiography.