To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Primary antibody reactivity:Mouse
- Description:Mouse/Rat Urocortin 3 ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-008
Mouse Rat Urocortin 3 ELISA determines urocortin 3 in plasma, serum and brain tissue extracts.
- Dynamic Range: 0.41 - 100 ng/mL
- Sensitivity: 0.41 ng/mL
- Incubation: 21 hours
Urocortin 3 (Ucn3) or stresscopin (SCP) is a new member of the corticotropin-releasing factor (CRF) peptide family identified in the mouse and human. The CRF family of neuropeptides includes mammalian peptides CRF, urocortin 1(Ucn1) and urocortin 2 (Ucn2) or stress-related peptide (SRP), as well as piscine urotensin 1 and frog sauvagine. Mouse and human Ucn3 share 90% identity in the 38-aa putative mature peptide.