Antibody to Omalizumab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-256
Supplier: Eagle Biosciences
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:
- Description:Antibody to Omalizumab ELISA Assay Kit
- Size:1 kit
- Environmentally Preferable:
- Cat. no.:103389-256
Specifications
About this item
Antibody to Omalizumab ELISA determines free antibodies to Omalizumab in serum and plasma samples.
- Dynamic Range: Cut-off
- Sensitivity: Cut-off
- Incubation: 2.5 hours
The Eagle Biosciences Antibody to Omalizumab is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Omalizumab in serum and plasma samples. The Antibody to Omalizumab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.