Order Entry
Canada
ContactUsLinkComponent
Antibody to Rituxinab ELISA Assay Kit, Eagle Biosciences
Antibody to Rituxinab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-250
Supplier:  Eagle Biosciences
CAS Number:  
Antibody to Rituxinab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-250
Supplier:  Eagle Biosciences
Supplier Number:  IG-BB106
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Host:
  • Primary antibody reactivity:
  • Description:
    Antibody to Rituxinab ELISA Assay Kit
  • Size:
    1 kit
  • Environmentally Preferable:
  • Cat. no.:
    103389-250

Specifications

About this item

Antibody to Rituximab ELISA to determine free antibodies to Rituximab in serum and plasma samples.

  • Dynamic Range: Cut-off
  • Sensitivity: Cut-off
  • Incubation: 2.5 hours

The Eagle Biosciences Antibody to Rituximab is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Rituximab (Rituxan®, Mabthera®) in serum and plasma samples. The Antibody to Rituximab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.