Antibody to Adalimumab ELISA Assay Kit, Eagle Biosciences
Catalog # 103389-244
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Description:Antibody to Adalimumab ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-244
Specifications
About this item
Antibodies to Adalimumab ELISA detects free antibodies to Adalimumab in serum and plasma samples.
- Dynamic Range: Cut-off
- Sensitivity: Cut-off
- Incubation: 2.5 hours
The Eagle Biosciences Antibody to Adalimumab ELISA Assay Kit is an enzyme immunoassay for the semi-quantitative determination of free antibodies to Adalimumab in serum and plasma samples. The Antibody to Adalimumab ELISA Assay Kit is for research use only and not to be used in diagnostic procedures.