Cetuximab (mAB-based) ELISA Assay Kit, Eagle Biosciences
103389-238
:
- Description:Cetuximab (mAB-based) ELISA Assay Kit
- Size:1 kit
- Cat. no.:103389-238
Cetuximab mAb-based ELISA precisely measures free Cetuximab in serum and plasma.
- Dynamic Range: 6 - 200 ng/mL
- Sensitivity: 2 ng/ml
- Incubation: 2 hours
The drug Cetuximab (trade name Erbitux®) is a human-mouse chimeric immunoglobulin (Ig) G1 monoclonal antibody (mAb), selectively directed against the epidermal growth factor receptor (EGFR), also known as HER1 or ErbB1. Since its approval by Food and Drug Administration for cancer treatment in 2004, Cetuximab became widely used in the treatment of colorectal and head and neck cancers. The ImmunoGuide Cetuximab ELISA (mAb-based) kit can be efficiently used for measuring free Cetuximab levels in serum and plasma.