To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Host:
- Primary antibody reactivity:Mouse
- Description:Mouse / Rat Noradrenaline (Norepinephrine) ELISA
- Size:1 kit
- Environmentally Preferable:
- Cat. no.:103389-150
Mouse/Rat Noradrenaline ELISA detects noradrenaline in biological samples
- Dynamic Range: 0.05 - 25 ng/mL
- Sensitivity: 1 pg/mL
- Incubation: Overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.
For Research Use Only