TNF alpha Neutralization Rate/Adalimumab ELISA Assay Kit, Eagle Biosciences
Catalog # 103388-996
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Target protein:TNF-alpha
- Description:TNF alpha Neutralization Rate/Adalimumab ELISA Assay Kit
- Size:1 kit
- Cat. no.:103388-996
Specifications
About this item
The recoveryELISA RTA is for research use only and not to be used in diagnostic procedures.
- Dynamic Range: Adalimumab: 0.7 – 30 µg/mL; TNF alpha in sample without Adalimumab: 4-400ng/ml;TNF alpha in sample with Adalimumab: 4-160ng/ml
- Sensitivity: Adalimumab: 0.7 µg/mL; TNF alpha: 4ng/ml
- Incubation: overnight
Tumour necrosis factor (TNFα) is a signalling substance of the immune system which can cause, among other things, the formation of inflammatory mediators in cells. In inflammatory diseases such as rheumatoid arthritis, there is overproduction of TNFα, which can be inhibited by using therapeutic antibodies.