Noradrenaline (Norepinephrine) High Sensitive ELISA, Eagle Biosciences
Catalog # 103389-148
Supplier: Eagle Biosciences
CAS Number:
Specifications
- Description:Noradrenaline (Norepinephrine) High Sensitive ELISA
- Size:1 kit
- Cat. no.:103389-148
Specifications
About this item
Noradrenaline High Sensitive ELISA detects noradrenaline in biological samples.
- Dynamic Range: 0.05 - 25 ng/mL
- Sensitivity: 1 pg/mL
- Incubation: overnight
Catecholamine is the name of a group of aromatic amines (noradrenaline, adrenaline, dopamine, and their derivatives) which act as hormones and neurotransmitter, respectively. Adrenaline and noradrenaline are formed from dopamine. They act on the cardiac musculature and the metabolism (adrenaline) as well as on the peripheral circulation (noradrenaline) and help the body to cope with acute and chronic stress.