Electrolyte Solution, L5014, SI Analytics
L5014 Electrolyte Solution, 250 mL, Bottle
97042-456
Supplier #: 285138324
Acetic acid saturated with lithium chloride.
- Pack type:Bottle
- Description:L5014 Electrolyte Solution
- Size:250 mL