Expres² TR Transfection Reagent
Catalog # 77705-788
Supplier: EXPRESSION SYSTEMS LLC
New Product
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:
- Cat. no.:77705-788
Specifications
About this item
Expres² TR transfection reagent, formulated and optimized for insect cell culture.
- Formulation works seamlessly with transfection medium
- Optimized for use in insect cell culture
- Consistently high transfection rates
- Increases virus vector production and transient expression
It is a cationic lipid formulation designed for the co-transfection or transfection step in baculovirus vector production.
Delivery: Priority overnight.