Czapek Dox Agar, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS CA
Certificates
About this item
Accommodates deeper fills. Used for the cultivation of saprophytic fungi and soil bacteria capable of utilizing inorganic nitrogen.
- Neutral pH
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.