Order Entry
Canada
ContactUsLinkComponent
HardyVal™ MTK (Multiple Technician Kit)
HardyVal™ MTK (Multiple Technician Kit)
Catalog # CA76102-166
CAS Number:  
undefined
HardyVal™ MTK (Multiple Technician Kit)
Catalog # CA76102-166
Supplier Number:  HVMTK
CAS Number:  

Specifications

  • Description:
    Multi-tech kit
  • Size:
    5 tests per kit
  • Cat. no.:
    CA76102-166
  • Application:
    Compounding pharmacy

Specifications

About this item

HardyVal™ MTK (Multiple Technician Verification Kit) is recommended for verification of personal aseptic technique for low and medium complexity levels within a sterile compounding pharmacy facility or other cleanroom application.

Each kit contains enough media to perform aseptic technique verification for up to five technicians. Product is quality control tested for growth promotion, pH, and sterility.

Delivery: Each kit contains 5 ampoules of TSB, 3 ml (non-toxic red dye is added to the TSB to easily visualize the sample being transferred), 5 vials of TSB 20 ml serum needle-port septum vial, 5 bags of TSB 100 ml media bag.