Order Entry
Canada
ContactUsLinkComponent
Accessories for Fastprep-96™ Sample Preparation Instrument, MP Biomedicals
Accessories for Fastprep-96™ Sample Preparation Instrument, MP Biomedicals
Catalog # CA76045-816
Supplier:  MP Biomedicals
Accessories for Fastprep-96™ Sample Preparation Instrument, MP Biomedicals
Catalog # CA76045-816
Supplier:  MP Biomedicals
Supplier Number:  116010570
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    QuickFlex™ Adapter for FastPrep-96™
  • Cat. no.:
    CA76045-816
  • Sample Size:
    96 x 2 mL

Specifications

About this item

High-throughput adapter for the FastPrep-96™ that holds 96 x 2 mL Lysing Matrix Tubes.

  • Ergonomic design
  • Easy loading of 2mL tubes
  • Stands stable on the benchtop
  • Commonly used as a tube rack

With the FastPrep-96 Instrument, QuickFlex adapter and optimized Lysing Matrices, consistent reproducible results are obtained.

QuickFlex™ 96 x 2 mL Adapter for FastPrep-96™ is a large sample volume adapter is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20 °C or -80 °C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.

Ideally suited for high-throughput applications, up to 96 samples can be homogenized simultaneously. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at –20°C or –80°C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.

Use with FastPrep-96, MP Bio Lysing Matrix Tubes and/or FastPrep purification kits to lyse, grind or homogenize samples at ambient temperatures, for the extraction and purification of nucliec acids, proteins, metabolites and other small molecules and more.