Order Entry
Canada
ContactUsLinkComponent
Accessories for Fastprep Sample Preparation Instruments BigPrep™, MP Biomedicals
Accessories for Fastprep Sample Preparation Instruments BigPrep™, MP Biomedicals
  CA76045-840
Accessories for Fastprep Sample Preparation Instruments BigPrep™, MP Biomedicals
  CA76045-840
 :  116002525
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:

  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address

Avantor will not lift restrictions for residential shipping addresses.

 

  • Description:
    BigPrep™ Adapter,2×50 ml
  • Cat. no.:
    CA76045-840

 

 

BigPrep™ 2 x 50 mL Adapter is a large-scale adapter that holds 2 x 50 mL Lysing Matrix tubes.

  • Wide range of disposable 50mL Lysing Matrix tubes
  • Thorough homogenization of any sample type in seconds
  • Avoids cross contamination typical of traditional homogenization methods

With the FastPrep-24 or FastPrep-24 5G Instruments, BigPrep adapter and optimized Lysing Matrices, consistent reproducible results are obtained.

BigPrep™ 2 x 50 mL Adapter is a large sample volume adapter that is ideally suited for DNA and RNA isolation, enzyme isolation and protein production, natural products isolation, food preparation for quality analysis, biopharma manufacturing, and forensic applications. Additionally, frozen samples in Lysing Matrix tubes loaded in the adapter and stored at -20 °C or -80 °C are ready to be immediately processed with minimal hands-on manipulation, preventing degradation of cellular components by endogenous enzymes.

Use with FastPrep-24, FastPrep-24 5G, or FastPrep-96 (with ConeFlex Legacy Adapter), MP Bio Lysing Matrix Tubes and/or FastPrep purification kits to lyse, grind or homogenize samples at ambient temperatures, for the extraction and purification of nucliec acids, proteins, metabolites and other small molecules and more.