ContactUsLinkComponent
SP Bel-Art Silicone-Free Lens Cleaning Tissues, Bel-Art Products, a part of SP
Catalog # 490005-582
undefined
SP Bel-Art Silicone-Free Lens Cleaning Tissues, Bel-Art Products, a part of SP
Catalog # 490005-582
Supplier Number:  F24841-0000
Specifications
  • Description:
    Silicone-Free Lens Cleaning Tissues
  • Cat. no.:
    490005-582
  • Packaging:
    150 Sheets/Pack
  • W×L:
    10.8 x 11.4 cm (4.25 x 4.5")
  • Supplier No.:
    F24841-0000
Specifications
About this item
Use these high-quality, lint-free tissues to quickly and easily clean glass and plastic lenses.
  • Tissues do not contain silicone
  • Will not scratch glass or plastic lenses
  • 150 sheets packaged in a pop-up dispenser pack