Order Entry
Canada
ContactUsLinkComponent
 
Marble Table
  CA11276-998
Marble Table
  CA11276-998
 :  63053976

 

  • Description:
    Marble table
  • Height:
    31"
  • Environmentally Preferable:
  • Cat. no.:
    CA11276-998
  • W×D:
    35×24"
  • Supplier No.:
    63053976

 

 

When working with balances, tihs marble table provides maximum stability and eliminates vibration.

  • Marble top thickness: 3"
  • Approximate shipping weight: 660 lbs.