ContactUsLinkComponent
Amyloid Stain Kit, Congo Red, Polysciences
Amyloid Stain Kit, Congo Red, Polysciences
Catalog # 75805-950
Supplier:  Polysciences Inc
CAS Number:  
Amyloid Stain Kit, Congo Red, Polysciences
Catalog # 75805-950
Supplier:  Polysciences Inc
Supplier Number:  24614-1
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Amyloid Stain Kit, Stains up to 125 slides
  • Size:
    1 Kit
  • Cat. no.:
    75805-950
  • Supplier No.:
    24614-1
Specifications
About this item
Amyloid stain kit used in the detection of amyloid in formalin fixed paraffin embedded (FFPE) tissue sections as well as frozen tissue.
  • Recommended section thickness is 10 microns
Delivery: Kit contains 250 mL bottle of each, congo red solution A, alkaline alcohol solution B, Gill's hematoxylin #3 Solution C.