Mouse Recombinant Leptin
1 mg
89143-980
: 201-035-M001
Produced in E. coli. Homologous to mouse serum leptin.
- Size:1 mg
- Cat. No.:89143-980
- Reconstitution instructions:Reconstitute with sterile water or 0.4% sodium bicarbonate, pH 8-9. Do not reconstitute to less than 0.1mg/ml. Further dilutions should be made with medium containing a carrier protein.
- Protein/peptide type:Recombinant
- Endotoxin content:<0.1EU/µg purified protein.
- Protein/peptide name:Leptin
- Formulation:Lyophilized from 0.0045mM sodium bicarbonate.
- Purity:>98% (SDS-PAGE)
- Species:Mouse
- Biological activity:Induces proliferation of BAF/3 cells stably transfected with the long form of human leptin receptor.
- Molecular weight:~16kDa.
- CAS No.:181030-10-4



