Anti-TL1A/TNFSF15 Rabbit Monoclonal Antibody (DyLight 550) [clone: 2116C]
Catalog # 103291-590
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:TL1A/TNFSF15
- Clonality:Monoclonal
- Clone:2116C
- Conjugation:DyLight® 550
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:9966
- Antigen synonyms:TL1A|VEGI192A|tumor necrosis factor ligand superfamily member 15|TNF superfamily ligand TL1A|member 15|tumor necrosis factor (ligand) superfamily|TL1Vascular endothelial cell growth inhibitor|VEGITNF ligand-related molecule 1|MGC129934|MGC129935|vascular endothelial growth inhibitor-192A
- Storage buffer:50mM Sodium Borate
- Storage temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human TL1A protein (amino acids 72-251) [UniProt O95150]
- Purification:Protein A or G purified
- Cat. no.:103291-590
- Supplier No.:MAB7442R
Specifications
About this item
The TL1A / TNFSF15 Antibody (2116C) [DyLight 550] from Novus Biologicals is a rabbit monoclonal antibody to TL1A / TNFSF15. This antibody reacts with human. The TL1A / TNFSF15 Antibody (2116C) [DyLight 550] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: TL1A/TNFSF15
Clonality: Monoclonal
Clone: 2116C
Conjugation: DyLight 550
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human