Anti-TBK1 Mouse Monoclonal Antibody (Biotin) [clone: 108A429]
Catalog # 103361-212
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:TBK1
- Clonality:Monoclonal
- Clone:108A429
- Conjugation:Biotin
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:29110
- Antigen synonyms:EC 2.7.11.1|NAKserine/threonine-protein kinase TBK1|NF-kappa-B-activating kinase|FLJ11330|EC 2.7.11|TBK 1|NAK serine/threonine-protein kinase TBK 1|T2K|TANK-binding kinase 1NF-kB-activating kinase
- Storage buffer:PBS
- Storage temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide corresponding to amino acids 563-577 (YYQFKKDKAERRLAYC) of human TBK1 was used as immunogen for this antibody (NP_037386).
- Purification:Protein G purified
- Cat. no.:103361-212
Specifications
About this item
The TBK1 Antibody (108A429) [Biotin] from Novus Biologicals is a mouse monoclonal antibody to TBK1. This antibody reacts with human, mouse, rat, bovine, canine. The TBK1 Antibody (108A429) [Biotin] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: TBK1
Clonality: Monoclonal
Clone: 108A429
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Bovine, Canine