Anti-SNX26 Goat Polyclonal Antibody
Catalog # 103367-962
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:SNX26
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 μg
- Gene ID:115703
- Antigen synonyms:Tc10/CDC42 GTPase-activating protein|Rho GTPase activating protein 33|Sorting nexin-26|rho GTPase-activating protein 33|sorting nexin 26|FLJ39019|neurite outgrowth multiadaptor RhoGAP protein|NOMA-GAP|Rho-type GTPase-activating protein 33|TCGAPSNX26
- Storage buffer:0.5 mg/ml Tris (pH 7.3) and 0.5% BSA
- Storage temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Peptide with sequence SWSLHSEGQTRSYC corresponding to C-Terminus according to NP_443180.2.
- Tested applications:, Peptide
- Purification:Immunogen affinity purified
- Cat. no.:103367-962
Specifications
About this item
The SNX26 Antibody from Novus Biologicals is a goat polyclonal antibody to SNX26. This antibody reacts with human. The SNX26 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Type: Primary
Antigen: SNX26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human