Anti-PRMT6 Rabbit Polyclonal Antibody
Catalog # 103360-888
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:PRMT6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:0.05 ml
- Gene ID:55170
- Antigen synonyms:FLJ10559|HMT1 hnRNP methyltransferase-like 6|Histone-arginine N-methyltransferase PRMT6|EC 2.1.1.-|HRMT1L6|Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6|HMT1 hnRNP methyltransferase-like 6 (S. cerevisiae)|protein arginine N-methyltransferase 6|protein arginine methyltransferase 6|FLJ51477|EC 2.1.1.125
- Storage buffer:Whole antisera with PBS and 0.05% BSA
- Storage temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:This antibody was developed a synthetic peptide corresponding to amino acids 23-43 (CEEDGAEREAALERPRRTKRER) of human PRMT6.
- Purification:Unpurified
- Cat. no.:103360-888
Specifications
About this item
The PRMT6 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PRMT6. This antibody reacts with human, mouse. The PRMT6 Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: PRMT6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse