Anti-nNOS Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 60718]
Catalog # 103289-626
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:nNOS
- Clonality:Monoclonal
- Clone:60718
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:4842
- Antigen synonyms:N-NOS|Peptidyl-cysteine S-nitrosylase NOS1|Constitutive NOS|bNOS|EC 1.14.13.39|nNOSNOS type I|brain|NOS|IHPS1|nitric oxide synthase 1 (neuronal)|Neuronal NOS
- Storage buffer:PBS
- Storage temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human nNOS protein (amino acids 218-1434) [UniProt P29475]
- Purification:Protein A or G purified
- Cat. no.:103289-626
- Supplier No.:MAB2416H
Specifications
About this item
The nNOS Antibody (60718) [HRP] from Novus Biologicals is a mouse monoclonal antibody to nNOS. This antibody reacts with human, mouse, rat. The nNOS Antibody (60718) [HRP] has been validated for the following applications: Western Blot.
Type: Primary
Antigen: nNOS
Clonality: Monoclonal
Clone: 60718
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat