ContactUsLinkComponent
Screw Cap Sample Tubes, Globe Scientific
Screw Cap Sample Tubes, Globe Scientific
Catalog #: 10022-134
Supplier:  Globe Scientific
TRANSPORTTUBE,4ML,PP,SELF-STAND,SCREWCAP
Screw Cap Sample Tubes, Globe Scientific
Catalog #: 10022-134
Supplier:  Globe Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Capacity:
    4 ml
  • Description:
    Transport Tube with Separate Natural Screw Cap
  • Material:
    PP
  • Supplier No.:
    6144
Specifications
About this item
These tubes and caps have been designed for the storage and transportation of biological material. They are manufactured from non-toxic polypropylene and offer strength and clarity.
  • Polypropylene screw caps provide same coefficient of expansion as the tubes
  • Leakproof seal even at changing temperatures
The vial has external threads, which provide a smooth and uniform inner surface and reduces the risk of contamination.
The caps feature a long skirt and a super-fast thread design that allows them to be removed or sealed with a single turn.
Customers Who Bought This Also Bought