Anti-DNMT1 Mouse Monoclonal Antibody (DyLight 350) [clone: 60B1220.1]
103359-620
- Antibody type:Primary
- Antigen symbol:DNMT1
- Clonality:Monoclonal
- Clone:60B1220.1
- Conjugation:DyLight® 350
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Human,Rat,Mouse
- Western blot:Yes
- Size:100 μl
- Gene ID:1786
- Antigen synonyms:DNA methyltransferase HsaI|CXXC9CXXC finger protein 9|DNMT|m.HsaI|DNA (cytosine-5-)-methyltransferase 1|AIM|DNA MTase HsaI|FLJ16293|DNA (cytosine-5)-methyltransferase 1|EC 2.1.1.37|CXXC-type zinc finger protein 9|Dnmt1|MCMTMGC104992|DNA methyltransferase 1
- Storage buffer:50mM Sodium Borate
- Storage temperature:Store at 4C in the dark.
- Immunogen:This antibody was raised against a synthetic peptide corresponding to amino acids 637-650 (EKDDREDKENAFKR) of human Dnmt1 (Genbank Accession No. NP_001370).
- Purification:Protein G purified
- Cat. no.:103359-620
The DNMT1 Antibody (60B1220.1) [DyLight 350] from Novus Biologicals is a mouse monoclonal antibody to DNMT1. This antibody reacts with human, mouse, rat, porcine. The DNMT1 Antibody (60B1220.1) [DyLight 350] has been validated for the following applications: Western Blot, Flow Cytometry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Type: Primary
Antigen: DNMT1
Clonality: Monoclonal
Clone: 60B1220.1
Conjugation: DyLight 350
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat, Porcine