Anti-DERP1 Human Monoclonal Antibody [clone: H.AA.IV7A3]
Catalog # 103318-800
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:DERP1
- Clonality:Monoclonal
- Clone:H.AA.IV7A3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgG1 kappa
- Reactivity:Insect
- Size:100 μg
- Antigen synonyms:DERP-1|Der p 1 allergen
- Storage buffer:Tris-NaCl (pH 8.0)
- Storage temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:Derp-1 was used as immunogen.
- Purification:Ion exchange chromatography
- Cat. no.:103318-800
- Supplier No.:DDX0211P-100
Specifications
About this item
The DERP1 Antibody (H.AA.IV7A3) from Novus Biologicals is a human monoclonal antibody to DERP1. This antibody reacts with insect, other. The DERP1 Antibody (H.AA.IV7A3) has been validated for the following applications: ELISA, ELISA Capture (Matched Antibody Pair).
Type: Primary
Antigen: DERP1
Clonality: Monoclonal
Clone: H.AA.IV7A3
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 Kappa
Reactivity: Insect, Other