Anti-CCNB1 Rabbit Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2061A]
Catalog # 103290-538
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Cyclin B1
- Antigen symbol:CCNB1
- Clonality:Monoclonal
- Clone:2061A
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:891
- Antigen synonyms:cyclin B1|G2/mitotic-specific cyclin-B1|G2/mitotic-specific cyclin B1|CCNB
- Storage buffer:PBS
- Storage temperature:Store at 4C in the dark.
- Immunogen:Partial recombinant human Cyclin-B1 protein (amino acids 1-91) [UniProt P14635]
- Purification:Protein A or G purified
- Cat. no.:103290-538
- Supplier No.:MAB6000F
Specifications
About this item
The Cyclin B1 Antibody (2061A) [FITC] from Novus Biologicals is a rabbit monoclonal antibody to Cyclin B1. This antibody reacts with human. The Cyclin B1 Antibody (2061A) [FITC] has been validated for the following applications: Western Blot, Flow Cytometry.
Type: Primary
Antigen: CCNB1
Clonality: Monoclonal
Clone: 2061A
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human