Order Entry
Canada
ContactUsLinkComponent
Unifrost™ Microscope Slides, Azer Scientific
Unifrost™ Microscope Slides, Azer Scientific
Catalogue # 89033-048
Fournisseur:  Azer Scientific Inc.
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise
undefined
Unifrost™ Microscope Slides, Azer Scientific
Catalogue # 89033-048
Fournisseur:  Azer Scientific Inc.
Supplier Number:  EMS200U
Product available on GSA Advantage®
Minority or Woman-Owned Business Enterprise

Spécifications

  • Color:
    Gold
  • Description:
    Frosted slides
  • L×W:
    75×25 mm
  • Thickness:
    1.0 mm
  • Cat. no.:
    89033-048
  • Packaging:
    72/Box, 20 Boxes/Case
  • Supplier No.:
    EMS200U

Spécifications

À propos de cet article

Manufactured from high-quality white German glass, with a single frosted end.

  • Single frosted end to help prevent sticking
  • Colored end helps with quick identification and organization

All slides have single side, color frosted ends which helps to avoid sticking and jamming problems that can occur with other slides. The color frosted end is resistant to all common chemicals that are utilized in the laboratory.