Anti-Avian Flu Nucleoprotein Rabbit Polyclonal Antibody
Catalog # 103360-434
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Avian Flu Nucleoprotein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Birds,Chicken
- Size:0.025 mg
- Antigen synonyms:Nucleoprotein|NP
- Storage buffer:PBS containing 0.05% BSA
- Storage temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:A synthetic peptide corresponding to amino acids 428-441 (AFTGNTEGRTSDMR) of avian flu nucleoprotein were used as the immunogen, GenBank no ADC34563.1.
- Purification:Protein G purified
- Cat. no.:103360-434
Specifications
About this item
The Avian Flu Nucleoprotein Antibody from Novus Biologicals is a rabbit polyclonal antibody to Avian Flu Nucleoprotein. This antibody reacts with avian, chicken. The Avian Flu Nucleoprotein Antibody has been validated for the following applications: ELISA, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: Avian Flu Nucleoprotein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Avian, Chicken