Anti-CD21 Mouse Monoclonal Antibody [clone: 7D6]
103318-704
- Antibody type:Primary
- Antigen symbol:CD21
- Clonality:Monoclonal
- Clone:7D6
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 μg
- Gene ID:1380
- Antigen synonyms:Complement C3d receptor|CD21 antigen|Epstein-Barr virus receptor|complement component (3d/Epstein Barr virus) receptor 2|EBV receptor|CD21|C3DRSLEB9|complement receptor type 2|Cr2
- Storage buffer:Tris-NaCl (pH 8.0)
- Storage temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:CD21 long isoform was used as immunogen.
- Purification:Ion exchange chromatography
- Cat. no.:103318-704
- Supplier No.:DDX0120P-100
The CD21 Antibody (7D6) from Novus Biologicals is a mouse monoclonal antibody to CD21. This antibody reacts with human. The CD21 Antibody (7D6) has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Frozen.
Type: Primary
Antigen: CD21
Clonality: Monoclonal
Clone: 7D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human