Anti-BST2 Rat Monoclonal Antibody [clone: 120G8.04]
Catalog # 103318-936
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:BST2
- Clonality:Monoclonal
- Clone:120G8.04
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Size:100 μg
- Neutralisation:Yes
- Gene ID:684
- Antigen synonyms:bone marrow stromal cell antigen 2|CD317|HM1.24 antigen|bone marrow stromal antigen 2|TETHERIN|NPC-A-7|BST-2|PDCA-1|CD317 antigen
- Storage buffer:Tris-NaCl (pH 8.0)
- Storage temperature:Store at -20C. Avoid freeze-thaw cycles.
- Concentration:0.5 mg/ml
- Immunogen:mouse plasmatocytoid DCs (pDCs)
- Purification:Ion exchange chromatography
- Cat. no.:103318-936
- Supplier No.:DDX0390P-100
Specifications
About this item
The BST2 Antibody (120G8.04) from Novus Biologicals is a rat monoclonal antibody to BST2. This antibody reacts with mouse. The BST2 Antibody (120G8.04) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Blocking / Neutralizing, In vivo assay.
In vivo assay
Type: Primary
Antigen: BST2
Clonality: Monoclonal
Clone: 120G8.04
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 Kappa
Reactivity: Mouse