Anti-BP1 Rabbit Polyclonal Antibody (Alexa Fluor® 700)
103358-146
- Antibody type:Primary
- Antigen symbol:BP1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 700
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:100 μl
- Gene ID:1748
- Antigen synonyms:homeobox protein DLX-4|DLX8DLX9|BP1distal-less homeo box 7|distal-less homeobox 4|DLX7distal-less homeo box 9|Homeobox protein DLX-7|Beta protein 1|Homeobox protein DLX-8
- Storage buffer:50mM Sodium Borate
- Storage temperature:Store at 4C in the dark.
- Immunogen:A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProt# Q92988].
- Purification:Immunogen affinity purified
- Cat. no.:103358-146
The BP1 Antibody [Alexa Fluor« 700] from Novus Biologicals is a rabbit polyclonal antibody to BP1. This antibody reacts with human. The BP1 Antibody [Alexa Fluor« 700] has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Type: Primary
Antigen: BP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human