Anti-Anthrax LF Rabbit Polyclonal Antibody
103360-984
- Antibody type:Primary
- Antigen name:Anthrax Lethal Factor
- Antigen symbol:Anthrax LF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Size:0.05 ml
- Antigen synonyms:Anthrax lethal factor|Anthrax lethal toxin endopeptidase component|B.anthracis lethal factor|LF|Anthrax LF|bacillus anthracis lethal factor|Lef
- Storage buffer:Whole antisera with PBS and 0.2% Gelatin
- Storage temperature:Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
- Immunogen:The antibody was developed by immunizing rabbits with synthetic peptides corresponding to amino acids 47-65 of the Anthrax LF protein (approx. 90 kD).
- Purification:Unpurified
- Cat. no.:103360-984
The Anthrax LF Antibody from Novus Biologicals is a rabbit polyclonal antibody to Anthrax LF. This antibody reacts with human. The Anthrax LF Antibody has been validated for the following applications: Western Blot.
Type: Primary
Antigen: Anthrax LF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human