You searched for: Syringe Filters
Syringe filters are attached to the end of a syringe to remove particles from a sample prior to analysis. Filtering liquids, the single-use devices force liquids through the filter either when fluids are initially drawn or delivered. Circular shaped in diameters that fit common syringe sizes, the cartridges remove fragments that would otherwise damage narrow bore equipment. Certified for low levels of UV-absorbing extractables, the syringe filters purify liquids for reproducible results.
VWR® Syringe Filters
Supplier: VWR International
Typical applications are clarification, sterile filtration, sample preparation and sterile venting.
Expand 40 Items
J.T.Baker®, Syringe Filters
Supplier: Avantor
These syringe filters are specifically designed for chromatography sample preparation applications and optimized to provide the most consistent results with less extractables.
Expand 92 Items
Acrodisc® Syringe Filters, 13 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These 13 mm syringe filters offer an available prefiltration layer and wide choice of filter media.
Expand 25 Items
ValuPrep Syringe Filters
Supplier: Cytiva
ValuPrep syringe filters offer the confidence of Pall membranes in economic 1000 packs.
Expand 19 Items
Acrodisc® Ion Chromatography Syringe Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These syringe filters are specifically designed for ion chromatography applications and optimized to provide the most consistent results when analyzing ionic species.
Expand 10 Items
Acrodisc® Syringe Filters, DMSO-Safe, Sterile, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These 25mm filters are ideal for the sterilization of media used for cell cryopreservation.
Expand 1 Items
Acrodisc® Syringe Filters for Prefiltration, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
For prefiltering heavily contaminated aqueous or solvent based samples.
Expand 17 Items
Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
Expand 31 Items
Acrodisc® Syringe Filters, 4 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These syringe filters are ideal for filtration of gas and organic solvents.
Expand 1 Items
Acrodisc® WBC (White Blood Cell) Syringe Filter, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The Acrodisc® WBC premium syringe filter is ideal for use in the capture and recovery of leukocytes and the separation of red blood cells. Acrodisc® WBC provides a simple and efficient method to isolate blood cells used in a variety of research applications including: Antigen specificity and modulation studies, immune receptor analysis, receptor protein studies, genotyping, gene expression, cell therapy.
Expand 2 Items
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The new Acrodisc® One and Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.
Expand 31 Items
Acrodisc® Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These 25 mm syringe filters offer an available prefiltration layer and wide choice of filter media.
Expand 20 Items
Acrodisc® Sterile Syringe Filters for Scale Up, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
These 25 mm filters are ideal for scale up.
Expand 5 Items
Acrodisc® Syringe Filters with Versapor® Membranes, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
For use in cell culture and other sensitive biological research.
Expand 3 Items
Acrodisc® Units with Mustang® E Membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Disposible, single-use units remove endotoxins in water, buffers, neutral sugars, and certain biological solutions.
Expand 1 Items
Nalgene® Syringe Filters, PTFE, 13 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Thermo Scientific™ Nalgene™ 13 mm Ø syringe Filters with PTFE membranes with filter solvents.
Expand 2 Items
Nalgene® Syringe Filters, Cellulose Acetate, 4 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
For cold sterilization of aqueous solutions, biological or immunological samples, and cell culture media components.
Expand 2 Items
Acrodisc® PSF Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
The new Acrodisc® PSF premium syringe filters combine the benefits of a multi-layered prefiltration device with an automation-certified filter housing.
Expand 12 Items
Nalgene® Syringe Filters, Surfactant-Free Cellulose Acetate and Cellulose Acetate, 25mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Filter sample volumes of 10 to 100 ml with these 25 mm Ø syringe filters.
Expand 8 Items
Nalgene® Syringe Filters, Polyethersulfone, 25mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Filter aqueous solutions and media with Thermo Scientific™ Nalgene™ 25 mm Ø Syringe Filters with PES (polyethersulfone) membrane.
Expand 2 Items
Nalgene® Syringe Filters, Nylon, 25mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
These 25 mm Ø Nylon syringe filters are ideal for filtering samples and mobile phases used in GC and HPLC.
Expand 4 Items
Target® Syringe Filters, Glass Microfiber, National Scientific™
Supplier: Thermo Scientific
Glass microfiber syringe filters are ideal for dissolution testing.
Expand 2 Items
Nalgene® Syringe Filters, Polyethersulfone, 13 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Thermo Scientific™ Nalgene™ 13 mm Ø syringe filters with PES (polyethersulfone) membrane with filter aqueous solutions and media.
Expand 1 Items
Syringe Filters, Color Coded, MicroSolv Technology
Supplier: MicroSolv Technology Corp
Color coded low volume syringe filters for HPLC, CE or dissolution testing.
Expand 54 Items
Nalgene® Syringe Filters, PTFE, 25 mm, Thermo Scientific
Supplier: Thermo Fisher Scientific
Filter solvents with Thermo Scientific™ Nalgene™ 25 mm Ø syringe filters with PTFE membranes.
Expand 2 Items
Target® Syringe Filters, (PES) Membrane, National Scientific™
Supplier: Thermo Scientific
These syringe filters are used for ion chromatography, tissue culture filtration and filtration of proteins and nucleic acids, as well as for filtration of high-temperature liquids.
Expand 2 Items
Target2™ Nylon Syringe Filters, Thermo Scientific
Supplier: NATIONAL SCIENTIFIC MX
Adding a syringe filtration step prior to injection not only helps to ensure more consistent and reliable results; it also helps protect delicate instruments and prolongs column life.
Expand 10 Items
Target® Syringe Filters, Cellulose Acetate, National Scientific™
Supplier: Thermo Scientific
Cellulose acetate membrane for biological or protein-based samples.
Expand 3 Items
Target® Syringe Filters, Regenerated Cellulose, National Scientific™
Supplier: Thermo Scientific
Hydrophilic regenerated cellulose membrane with low extractables and lowest nonspecific binding for biological applications or problem pharmaceutical samples.
Expand 2 Items
Corning® Plastic Syringe Filters, Corning
Supplier: Corning
Syringe filters are available with a choice of membrane type: Polyethersulfone (PES) for low protein binding and faster flow rates; surfactant-free cellulose acetate (SFCA), with or without prefilter, for lowest protein binding; Polytetrafluoroethylene (PTFE) for chemical resistance; regenerated cellulose (RC) – the best choice for DMSO compatibility; and nylon (NY) which is hydrophilic, surfactant-free and lowest extractable.