Order Entry
Canada
Orders LinkContactUsLinkComponent
Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer

Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer

Digi-Sense® 104A PL312 Variable- Time Output Controller 1440 Watts, 120 V, 50/60 Hz
Catalog # 76183-396
Supplier:   Antyila Scientific
Supplier #:  36225-91
Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer
Catalog # 76183-396
Supplier:  Antyila Scientific
Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer
Digi-Sense® Variable Time Temperature Controllers, Cole-Parmer

Digi-Sense® 104A PL312 Variable- Time Output Controller 1440 Watts, 120 V, 50/60 Hz

Price
$762.09
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Manually set power control dial from 5 to 100% to control the output wattage.

  • Simplify temperature control with repeating timed On/Off cycles
  • Pilot light on front lets users know when the load is heating

Ordering information: Includes 6 ft. attached power cord.

Specifications
  • Description:
    Digi-Sense® 104A PL312 Variable- Time Output Controller 1440 Watts
  • Electrical:
    120 V, 50/60 Hz
  • Cat. No.:
    76183-396
  • Display type:
    Reference knob, Low to High
  • Control type:
    Output 1: Proportional Voltage, Timer