HyFlex® 11-947 Cut, Abrasion, and Oil Resistant Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
Designed for fine tactile handling that requires very good protection against cuts.
- Good mechanical protection
- Microfinished nitrile coating
The HyFlex® 11-947 gloves offer excellent levels of cut, abrasion, and oil resistance in various dip geometries. The liner is made from Ansell INTERCEPT TECHNOLOGY® for excellent cut performance and comfort. ISO Cut Level B, along with FORTIX™ Technology are provided for increased abrasion resistance. The microfinished nitrile coating also provides excellent levels of grip and is resistant to residual oils and greases present on handled parts.
Certifications: Meets ANSI Cut Level A2 and ANSI Abrasion Level 4, ISO Cut Level B.
Specifications
- Catalog No:
- 75840-716
- 75840-718
- 75840-720
- 75840-722
- 75840-724
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- 3/4 Dipped
- 3/4 Dipped
- 3/4 Dipped
- 3/4 Dipped
- 3/4 Dipped
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist